Läs mer
Nefopam N-Oxide, TRC
Produktkod. 30483083
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30483083 10 mg
30472033 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30483083 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN38941510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 66091-32-5
Molekylformel C17H19NO2
Molekylvikt (g/mol) 269.34
Synonym 3,4,5,6-Tetrahydro-5-methyl-1-phenyl-1H-2,5-benzoxazocine 5-Oxide
IUPAC-namn 5-methyl-5-oxido-1-phenyl-1,3,4,6-tetrahydro-2,5-benzoxazocin-5-ium
LEDER C[N+]1([O-])CCOC(c2ccccc2)c3ccccc3C1
Specifikationer
Kemiskt namn eller material Nefopam N-Oxide
Kvantitet 10 mg
InChI formel InChI=1S/C17H19NO2/c1-18(19)11-12-20-17(14-7-3-2-4-8-14)16-10-6-5-9-15(16)13-18/h2-10,17H,11-13H2,1H3
Formel vikt 269.1416