missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK1 Recombinant Human Protein
NEK1 Recombinant Human Protein
Brand: Invitrogen PV4202
Additional Details : Weight : 0.05250kg
Description
NEK1 is a serine⁄threonine protein kinase, related to the NIMA cell cycle regulator, and is highly expressed in meiotic germ cells. The kinase may function early in the DNA damage response pathway. NEK1 may also participate in polycystic kidney disease and neural cell development.Specifications
NP_036356 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
85.3kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
NEK1 | |
NEK1 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 667μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 1-505 | |
590nmoles of phosphate transferred to myelin basic protein (MBP) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1.67μg/mL | |
GST Tag | |
Kinase and inhibitors | |
85% by SDS-PAGE |
0.34mg/mL | |
4750, 84634 | |
NEK1 Recombinant Human Protein | |
–80°C | |
1 Each | |
NEK1 Recombinant Human Protein | |
NEK | |
NEK1 Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
For Research Use Only. Not for use in diagnostic procedures.