missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NEK4 Recombinant Human Protein

NEK4 is a serine/threonine kinase.

Brand:  Invitrogen™ PR7180A

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15448533

  • 29100.00 SEK / 100µg
Estimated Shipment: 07-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NEK4 is involved in signal transduction and cellular communication. It is proposed to act exclusively upon threonine residues and is present in most primary carcinomas.

Specifications

Specifications

NP_003148
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol
100μg
RUO - research use only
NEK4, NRK2
Human
GST Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
122kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
NEK Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers