Learn More
Invitrogen™ NEK6 Recombinant Human Protein
NEK6 kinase is a member of the NEK family. NEK6 is a serine/threonine kinase that controls the initiation of mitosis and is required for the metaphase-anaphase transition.
Brand: Invitrogen™ A31512
Additional Details : Weight : 0.01000kg
Description
This purified recombinant full-length protein can be used in a number of different applications, but it has been specifically validated for use with our Z'-LYTE™ assay technology to identify novel kinase inhibitors. NEK6 is also available in our SelectScreen™ Kinase Profiling Service.
- Validated for use with the Z'-LYTE assay technology
- Available in multiple pack sizes for convenient use
- Also available in our SelectSceen Kinase Profiling Service
Specifications
NP_055212.2 | |
50mM Tris-HCl (pH 7.5), 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, and 25% Glycerol. | |
100μg | |
Full Length, Recombinant, Active | |
NEKs | |
Human | |
GST Tag |
Kinase Assay, Target and Lead Identification and Validation, Kinase Biology, Kinase Assay, Industrial and Applied Science, Pharma and Biopharma | |
62.4kDa | |
Store at -80°C. Upon first use, aliquot and store at -80°C to avoid multiple freeze/thaws. | |
SID6-1512 | |
Full Length | |
Recombinant | |
Insect |