missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Nelarabine
GREENER_CHOICE

Product Code. 18799916
Change view
Click to view available options
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
18799916 10 mg
18782469 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18799916 Supplier Tocris Bioscience™ Supplier No. 6359/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purine nuceloside analog; inhibits DNA synthesis

Nelarabine is a nucleoside analog. Metabolized by adenosine deaminase to active metabolite, arabinofuranosylguanine (ara-G). Inhibits DNA synthesis and induces apoptosis. Inhibits growth of human T cell lymphoblastic leukemia cells in vitro and in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 9-β-D-Arabinofuranosyl-6-methoxy-9H-purin-2-amine
CAS 121032-29-9
Analysprocentintervall >98%
Kvantitet 10 mg
Molekylformel C11H15N5O5
Formel vikt Observed MW: 297.27
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.