Läs mer
Nelarabine, Tocris Bioscience™
Purine nuceloside analog; inhibits DNA synthesis
Brand: Tocris Bioscience 6359/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Nelarabine is a nucleoside analog. Metabolized by adenosine deaminase to active metabolite, arabinofuranosylguanine (ara-G). Inhibits DNA synthesis and induces apoptosis. Inhibits growth of human T cell lymphoblastic leukemia cells in vitro and in vivo.Specifikationer
| 9-β-D-Arabinofuranosyl-6-methoxy-9H-purin-2-amine | |
| 121032-29-9 | |
| >98% | |
| 50 mg |