Läs mer
Neo Heliopan AP Sodium Salt, TRC
Produktkod. 30012825
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012825 100 mg
30005107 1 g
30006869 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012825 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN389945100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 226952-42-7
Molekylformel C20H14N4O12S4 xNa
Molekylvikt (g/mol) 653.6
Synonym 1H-Benzimidazole-5,7-disulfonic acid, 2,2'-(1,4-phenylene) Sodium Salt, 1H-Benzimidazole-4,6-disulfonic acid, 2,2'-(1,4-phenylene) Sodium Salt, Bisdisulizole Sodium, Bisimidazylate Sodium Salt, Sodium Bisdisulizole, Disodium Phenyldibenzimidazoletetrasulfonate Sodium Salt, Neo Heliopan Sodium Salt, Neo Heliopan APC Sodium Salt
IUPAC-namn 2,2'-(1,4-phenylene)bis(1H-benzo[d]imidazole-4,6-disulfonic acid), sodium salt
LEDER O=[S](O)(C1=CC([S](=O)(O)=O)=CC2=C1N=C(C3=CC=C(C4=NC(C([S](=O)(O)=O)=CC([S](=O)(O)=O)=C5)=C5N4)C=C3)N2)=O.Na
Specifikationer
Kemiskt namn eller material Neo Heliopan AP Sodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C20H14N4O12S4.Na/c25-37(26,27)11-5-13-17(15(7-11)39(31,32)33)23-19(21-13)9-1-2-10(4-3-9)20-22-14-6-12(38(28,29)30)8-16(18(14)24-20)40(34,35)36;/h1-8H,(H,21,23)(H,22,24)(H,25,26,27)(H,28,29,30)(H,31,32,33)(H,34,35,36);/q;+1
Formel vikt 651.931
Procent renhet >95%