Läs mer
Neodymium Gallium Oxide Substrate, 10 x 10 x 0.5mm, Polished One Side, 110 Orientation, Thermo Scientific™
Kvantitet:
1 Ea.
5 Ea.
Förpackningsstorlek:
Paket med 5
Varje
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Neodymium Gallium Oxide Substrate |
| Namnnotering | Polished One Side |
| Kvantitet | 1 Ea. |
| Molekylformel | NdGaO3 |
| MDL-nummer | MFCD00210638 |