missing translation for 'onlineSavingsMsg'
Learn More

Neodymium powder, -40 mesh, 99.8% (REO), Thermo Scientific Chemicals

Catalog Number p-7059211
Click to view available options
Quantity:
5 g
25 g
7440-00-8
Nd
144.24
MFCD00011130
QEFYFXOXNSNQGX-UHFFFAOYSA-N
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
23934
CHEBI:33372
neodymium
[Nd]
7440-00-8
Nd
144.24
MFCD00011130
QEFYFXOXNSNQGX-UHFFFAOYSA-N
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
23934
CHEBI:33372
neodymium
[Nd]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Color Gray to Silver
UN Number UN3089
Quantity 5 g
Percent Purity 99.9%
Mesh Size −40 Mesh
Odor Odorless
Packaging Ampule
Sensitivity Air and moisture sensitive
Form Powder
Assay (REO)
Chemical Name or Material Neodymium powder
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'