Läs mer
Neomenthyl Acetate, TRC
Produktkod. 30470360
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30470360 100 mg
1 options
Produktkod. 30470360 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN390220100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Neomenthyl Acetate
CAS 2230-87-7
Molekylformel C12H22O2
Kvantitet 100 mg
Synonym (±)-Neomenthyl Acetate, trans-Neomenthyl Acetate, (1S,2S,5R)-2-Isopropyl-5-methylcyclohexyl Acetate
InChI formel InChI=1S/C12H22O2/c1-8(2)11-6-5-9(3)7-12(11)14-10(4)13/h8-9,11-12H,5-7H2,1-4H3/t9-,11+,12+/m1/s1
LEDER C[C@H]1C[C@H](OC(C)=O)[C@H](C(C)C)CC1
IUPAC-namn (1S,2S,5R)-2-isopropyl-5-methylcyclohexyl acetate
Molekylvikt (g/mol) 198.3
Formel vikt 198.16