missing translation for 'onlineSavingsMsg'
Läs mer

Neomenthyl Acetate, TRC

Produktkod. 30477359
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30477359 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30477359 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN39022025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Neomenthyl Acetate
CAS 2230-87-7
Molekylformel C12H22O2
Kvantitet 25 mg
Synonym (±)-Neomenthyl Acetate, trans-Neomenthyl Acetate, (1S,2S,5R)-2-Isopropyl-5-methylcyclohexyl Acetate
InChI formel InChI=1S/C12H22O2/c1-8(2)11-6-5-9(3)7-12(11)14-10(4)13/h8-9,11-12H,5-7H2,1-4H3/t9-,11+,12+/m1/s1
LEDER C[C@H]1C[C@H](OC(C)=O)[C@H](C(C)C)CC1
IUPAC-namn (1S,2S,5R)-2-isopropyl-5-methylcyclohexyl acetate
Molekylvikt (g/mol) 198.3
Formel vikt 198.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.