Läs mer
Neomycin B Hexaacetate, TRC
Produktkod. 30483165
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30483165 5 mg
30474245 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30483165 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN3899905MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 119-04-0
Molekylformel C23 H46 N6 O13
Molekylvikt (g/mol) 614.64
Synonym O-2,6-diamino-2,6-dideoxy-alpha-D-glucopyranosyl-(1→4)-O-[O-2,6-diamino-2,6-dideoxy-beta-L-idopyranosyl-(1→3)-beta-D-ribofuranosyl-(1→5)]-2-deoxy-D-Streptamine, O-2,6-Diamino-2,6-dideoxy-beta-L-idopyranosyl-(1→3)-O-beta-D-ribofuranosyl-(1→5)-O-[2,6-diamino-2,6-dideoxy-alpha-D-glucopyranosyl-(1→4)]-2-deoxy-D-streptamine
IUPAC-namn (2R,3S,4R,5R,6R)-5-amino-2-(aminomethyl)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-2-[(2S,3R,4S,5R)-4-[(2R,3R,4R,5S,6S)-3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy-3-hydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy-3-hydroxycyclohexyl]oxyoxane-3,4-diol
LEDER NC[C@@H]1O[C@H](O[C@H]2[C@@H](O)[C@H](O[C@@H]3[C@@H](O)[C@H](N)C[C@H](N)[C@H]3O[C@H]4O[C@H](CN)[C@@H](O)[C@H](O)[C@H]4N)O[C@@H]2CO)[C@H](N)[C@@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material Neomycin B
Kvantitet 5 mg
InChI formel InChI=1S/C23H46N6O13/c24-2-7-13(32)15(34)10(28)21(37-7)40-18-6(27)1-5(26)12(31)20(18)42-23-17(36)19(9(4-30)39-23)41-22-11(29)16(35)14(33)8(3-25)38-22/h5-23,30-36H,1-4,24-29H2/t5-,6+,7-,8+,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23+/m1/s1
Formel vikt 614.3123