missing translation for 'onlineSavingsMsg'
Läs mer

Neopentylbenzene, 97+%

Produktkod. p-7020848
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 g
25 g
Förpackningsstorlek:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11459773 5 g 5g
11469773 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11459773 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer A12941.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 1007-26-7 | C11H16 | 148.249 g/mol

It is used in the preparation of α-Disulfones by Cobalt(III) Oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of α-Disulfones by Cobalt(III) Oxidation.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from ignition sources. Keep away from electrostatic charges.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1007-26-7
Molekylformel C11H16
Molekylvikt (g/mol) 148.249
MDL-nummer MFCD00038279
InChI-nyckel CJGXJKVMUHXVHL-UHFFFAOYSA-N
Synonym neopentylbenzene, benzene, 2,2-dimethylpropyl, neopentyl benzene, benzene, neopentyl, 2,2-dimethyl-1-propyl benzene, unii-7mhd03e509, 2,2-dimethylpropyl benzene, 1-neopentylbenzene, acmc-1bs8x
PubChem CID 13877
IUPAC-namn 2,2-dimethylpropylbenzene
LEDER CC(C)(C)CC1=CC=CC=C1

Specifikationer

Kemiskt namn eller material Neopentylbenzene
Densitet 0.859
Kokpunkt 184°C to 185°C
Flampunkt 50°C (122°F)
Brytningsindex 1.488
Kvantitet 5 g
UN-nummer UN3295
Beilstein 2037544
Löslighetsinformation Slightly soluble in water.
Formel vikt 148.25
Procent renhet ≥97%
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.