Learn More
Kartell™ Neoprene O-rings
Complete a filtration and vacuum desiccator with a spare o-ring. Kartell™ Neoprene O-rings form a seal with the transparent cover and polypropylene base of a filtration and vacuum desiccator.
39.00 SEK - 62.20 SEK
Specifications
Color Black
Product Type O-Ring
Material Neoprene
Quantity 1Each
View More Specs
Products 3
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
11969898
View Documents Promotion Details
Kartell™
0055700
For 8890A
62.20 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
12523357
View Documents Promotion Details
Kartell™
0055500
For 8890X
39.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
12034707
View Documents Promotion Details
Kartell™
0055600
For 230
60.40 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
Description
Description
  • Essectial component of a Filtration and Vacuum Desiccator
  • Spare O-ring
  • Spread a little grease on O-ring for assistance in establishing initial vacuum
Specifications
Specifications
Black
O-Ring
Neoprene
1Each