Läs mer
NEpsilon-(1-Carboxyethyl)-L-lysine(Mixture of Diastereomers), TRC
Produktkod. 30010417
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010417 10 mg
30011935 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010417 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17807010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5746-03-2
Molekylformel C9H18N2O4
Molekylvikt (g/mol) 218.25
Synonym Nepsilon-(1-Carboxyethyl)lysine, N6-(1-Carboxyethyl)-L-lysine, N6-(1'-Carboxyethyl)lysine, CEL;
LEDER CC(NCCCC[C@H](N)C(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material Nepsilon-(1-Carboxyethyl)-L-lysine(Mixture of Diastereomers)
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
Formel vikt 218.1267
Procent renhet >95%