Läs mer
NEpsilon-(1-Carboxymethyl)-L-lysine, TRC
Produktkod. 30011017
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011017 5 mg
30010292 1 mg
30008730 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011017 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1800755MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5746-04-3
Molekylformel C8 H16 N2 O4
Molekylvikt (g/mol) 204.22
Synonym L-Lysine, N6-(carboxymethyl)-, Lysine, N6-(carboxymethyl)-, L- (8CI), N6-(Carboxymethyl)-L-lysine, N6-(Carboxymethyl)lysine, Nε-(Carboxymethyl)lysine, ε-(Carboxymethyl)lysine
IUPAC-namn (2S)-2-amino-6-(carboxymethylamino)hexanoic acid
LEDER N[C@@H](CCCCNCC(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material Ne-(1-Carboxymethyl)-L-lysine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C8H16N2O4/c9-6(8(13)14)3-1-2-4-10-5-7(11)12/h6,10H,1-5,9H2,(H,11,12)(H,13,14)/t6-/m0/s1
Kvantitet 5 mg
Formel vikt 204.111
Procent renhet >95%