Läs mer
Nepsilon-Palmitoyl-L-glutamic Acid gamma-Succinimidyl-alpha-tert-butyl Ester, TRC
Produktkod. 30412324
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30412324 1 g
1 options
Produktkod. 30412324 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP1571001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Nepsilon-Palmitoyl-L-glutamic Acid gamma-Succinimidyl-alpha-tert-butyl Ester
CAS 204521-63-1
Molekylformel C29 H50 N2 O7
Kvantitet 1 g
Synonym Nε-Palmitoyl-Glu-γ-succinimidyl-α-tert-butyl Ester, (2S)-5-[(2,5-Dioxo-1-pyrrolidinyl)
InChI formel InChI=1S/C29H50N2O7/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-24(32)30-23(28(36)37-29(2,3)4)19-22-27(35)38-31-25(33)20-21-26(31)34/h23H,5-22H2,1-4H3,(H,30,32)/t23-/m0/s1
LEDER CCCCCCCCCCCCCCCC(=O)N[C@@H](CCC(=O)ON1C(=O)CCC1=O)C(=O)OC(C)(C)C
IUPAC-namn 1-O-tert-butyl 5-O-(2,5-dioxopyrrolidin-1-yl) (2S)-2-(hexadecanoylamino)pentanedioate
Molekylvikt (g/mol) 538.72
Formel vikt 538.3618