Läs mer
Neral, TRC
Produktkod. 30471001
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30471001 25 mg
30467368 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30471001 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN38291025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Neral
CAS 106-26-3
Anteckningar om renhetsgrad HPLC
Molekylformel C10H16O
Kvantitet 25 mg
Synonym (Z)-3,7-dimethylocta-2,6-dienal
InChI formel InChI=1S/C10H16O/c1-9(2)5-4-6-10(3)7-8-11/h5,7-8H,4,6H2,1-3H3/b10-7-
LEDER C(\CCC=C(C)C)(=C\C=O)/C
Molekylvikt (g/mol) 152.24