Learn More
Thermo Scientific™ NERL™ High Purity Water
Product Code. 15312208
Change view
Click to view available options
Quantity:
5 gal.
Unit Size:
Each
Product Code. Quantity Unit Size
15312208 5 gal. Each
1 options
  • For in vitro diagnostic and laboratory use only
  • Not for infusion, injection or irrigation
Product Code. 15312208 Supplier Thermo Scientific™ Supplier No. 9805
  • ONLINE EXCLUSIVE PRICE
    1555.00 SEK
    903.32 SEK / Paket med 1
    Save 651.68 SEK
    41% Av
Beräknad leverans: 17-03-2027
  • For in vitro diagnostic and laboratory use only
  • Not for infusion, injection or irrigation
Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ High Purity Water in place of deionized or distilled water.
Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ High Purity Water in place of deionized or distilled water. This product meets CAP/CLSI specifications for Clinical Laboratory Reagent Grade Water (CLRW) at time of manufacture.
Typically prepared at 18 megohm/cm specific resistance using reverse osmosis, mixed bed deionization, activated carbon filtration and final filtration to 0.2 micron, NERL high purity water is essentially free from organic and inorganic particulate and soluble contaminants.
Contains no preservatives
Ordering Information:
Certificate of analysis available for each case
TRUSTED_SUSTAINABILITY
Specifications
pH 5 to 7
Grade NERL
Quantity 5 gal.
General Purpose - For in vitro diagnostic or laboratory use only. Not for infusion, injection or irrigation.