Läs mer
Fisherbrand™ Nessler Cylinder
Produktkod. p-8002103 Handla allt Fisherbrand Produkter
Kapacitet (metrisk):
50 mL
100 mL
Förpackningsstorlek:
Paket med 2
Produktkod. 11932168
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Manufactured from soda lime glass. Fisherbrand™ Nessler Cylinders are ideal for colorimetric analysis.
  • For comparison of color and turbidity between liquids or solutions
  • Fixed values of 50mL or 100mL with 50mL graduation
  • Optically plane bottom provides maximum clarity for visual comparisons
  • Supplied in matched pairs to ensure uniformity in product for accuracy
TRUSTED_SUSTAINABILITY
Specifikationer
Tolerans ±0.8 mL
Material Glass
Bastyp Flat
Diameter (metrisk) Utsida 34 mm
Kapacitet (metrisk) 100 mL
Höjd (metrisk) 180 mm
Produkttyp Nessler Tube