Läs mer
Netobimin Sodium Salt, TRC
Produktkod. 17984525
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17984525 10 mg
17944825 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17984525 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN39090010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 88254-99-3
Molekylformel C14 H19 N4 O7 S2 . Na
Molekylvikt (g/mol) 442.44
Synonym Ethanesulfonic acid, 2-[[[(methoxycarbonyl)amino][[2-nitro-5-(propylthio)phenyl]amino]methylene]amino]-, monosodium salt (9CI)
IUPAC-namn sodium;2-[[(Z)-N-methoxycarbonyl-N'-(2-nitro-5-propylsulfanylphenyl)carbamimidoyl]amino]ethanesulfonate
LEDER [Na+].CCCSc1ccc(c(c1)\N=C(\NCCS(=O)(=O)[O-])/NC(=O)OC)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material Netobimin Sodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C14H20N4O7S2.Na/c1-3-7-26-10-4-5-12(18(20)21)11(9-10)16-13(17-14(19)25-2)15-6-8-27(22,23)24;/h4-5,9H,3,6-8H2,1-2H3,(H,22,23,24)(H2,15,16,17,19);/q;+1/p-1
Formel vikt 442.0593
Procent renhet >95%