Läs mer
Netupitant metabolite Netupitant N-oxide, MedChemExpress
Beskrivning
Netupitant N-oxide is the metabolite of Netupitant, which is a highly selective NK1 receptor antagonist.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Netupitant metabolite Netupitant N-oxide |
| CAS | 910808-11-6 |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C30H32F6N4O2 |
| Kvantitet | 1 mg |
| Synonym | Netupitant N-oxide |
| LEDER | CN(CCN1C2=CC(C3=CC=CC=C3C)=C(N(C)C(C(C)(C)C4=CC(C(F)(F)F)=CC(C(F)(F)F)=C4)=O)C=N2)(CC1)=O |
| Molekylvikt (g/mol) | 594.59 |
| Formel vikt | 594.59 |
| Kvalitet | Research |
| Visa mer |