Läs mer
STEMCELL Technologies NeuroCult™ Chemical Dissociation Kit (Mouse)
Produktkod. 17178281
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 sats
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17178281 1 sats
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 17178281 Leverantör STEMCELL Technologies Leverantörsnummer 05707
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Kit for chemical dissociation of mouse neurospheres
  • The NeuroCult™ Chemical Dissociation Kit is recommended for the non-mechanical and non-enzymatic dissociation of neurospheres derived from embryonic or adult mouse central nervous system tissue
  • A significantly higher viability and total cell number is observed after expansion, in comparison to neurospheres dissociated by trituration, and the functional properties of the cells are maintained
Specifikationer
Celltyp Neural Stem and Progenitor Cells
Kvantitet 1 sats