missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurogranin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10440-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Neurogranin is the human homolog of the neuron-specific rat RC3/neurogranin gene. This gene encodes a postsynaptic protein kinase substrate that binds calmodulin in the absence of calcium. The NRGN gene contains four exons and three introns. The exons 1 and 2 encode the protein and exons 3 and 4 contain untranslated sequences. It is suggested that the NRGN is a direct target for thyroid hormone in human brain, and that control of expression of this gene could underlay many of the consequences of hypothyroidism on mental states during development as well as in adult subjects.Specifications
Neurogranin | |
Polyclonal | |
Unconjugated | |
NRGN | |
hng, Neurogranin, Ng, NRGN, P17, RC3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4900, 64011, 64356 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P60761, Q04940, Q92686 | |
NRGN | |
Neurogranin Fusion Protein Ag0676 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |