missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Neuropeptide S, Human

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J66262.|D

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15437549

  • 2767.41 SEK / 0.50mg


Estimated Shipment
20-11-2020
Add to basket

Specifications

Specifications

2187.50
Human
Solid
RUO
Neuropeptide S, human
HPLC
0.5mg
Ser-Phe-Arg-Asn-Gly-Val-Gly-Thr-Gly-Met-Lys-Lys-Thr-Ser-Phe-Gln-Arg-Ala-Lys-Ser
-30° to -10°C
Solid
Videos
SDS Documents

For Research Use Only