Läs mer
Tocris Bioscience™ Neuropeptide S (Mouse)
Potent endogenous neuropeptide S receptor (NPSR) agonist
Brand: Tocris Bioscience™ 5857/1
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Neuropeptide S (Mouse) is a potent endogenous neuropeptide S receptor (NPSR) agonist (EC50 = 3 nM). Induces mobilization of intracellular Ca2+. Increases locomotor activity and wakefulness in mice. Also reduces anxiety-like behavior in mice.Specifikationer
| 412938-74-0 | |
| Additional Peptide Receptor Products | |
| 1 mg |
| Store at -20°C | |
| Observed M.W.: 2182.47 | |
| C93H156N34O27 |