Läs mer
Nevirapine Dimer, TRC
Product Code. 30468001
Change view
Click to view available options
Kvantitet:
10 mg
Product Code. Kvantitet
30468001 10 mg
1 options
Product Code. 30468001 Supplier Toronto Research Chemicals Supplier No. TRCN39129010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Nevirapine Dimer
CAS 1391054-30-0
Molekylformel C30H26N8O2
Kvantitet 10 mg
Synonym 11,11’-Dicyclopropyl-4,4’-dimethyl-5,5’,11,11’-tetrahydro-6H,6’H-9,9’-bidipyrido[3,2-b:2’,3’-e][1,4]diazepin-6,6’-dione, Nevirapine Impurity D, Nevirapine EP Impurity D
InChI formel InChI=1S/C30H26N8O2/c1-15-11-13-31-27-23(15)35-29(39)19-7-9-21(33-25(19)37(27)17-3-4-17)22-10-8-20-26(34-22)38(18-5-6-18)28-24(36-30(20)40)16(2)12-14-32-28/h7-14,17-18H,3-6H2,1-2H3,(H,35,39)(H,36,40)
LEDER CC1=C2C(=NC=C1)N(C3=C(C=CC(=N3)C4=NC5=C(C=C4)C(=O)NC6=C(C=CN=C6N5C7CC7)C)C(=O)N2)C8CC8
IUPAC-namn 11-cyclopropyl-9-(11-cyclopropyl-4-methyl-6-oxo-5H-dipyrido[4,2-a:2',3'-d][1,4]diazepin-9-yl)-4-methyl-5H-dipyrido[4,2-a:3',2'-e][1,4]diazepin-6-one
Molekylvikt (g/mol) 530.58
Formel vikt 530.2179