Läs mer
Nevirapine EP Impurity A, Mikromol™
Produktkod. 30461316
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30461316 4 x 25 mg
30052514 25 mg
30050082 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30461316 Leverantör Mikromol Leverantörsnummer MM1146.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Nevirapine EP Impurity A, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 11-Ethyl-4-methyl-5,11-dihydro-6H-dipyrido [3,2-b:2',3'-e][1,4]diazepin-6-one
CAS 133627-17-5
Analyt- eller komponentnamn 11-Ethyl-4-methyl-5,11-dihydro-6H-dipyrido [3,2-b:2',3'-e][1,4]diazepin-6-one
Frakt skick Room Temperature
Molekylformel C14 H14 N4 O
Certifikat och efterlevnad ISO 17025
Synonym Nevirapine Anhydrous Imp. A (EP), Nevirapine USP Related Compound A, Nevirapine USP RC A, 11-Ethyl-4-methyl-5,11-dihydro-6H-dipyrido [3,2-b:2',3'-e][1,4]diazepin-6-one, Nevirapine Related Compound A, Nevirapine Impurity A
InChI formel InChI=1S/C14H14N4O/c1-3-18-12-10(5-4-7-15-12)14(19)17-11-9(2)6-8-16-13(11)18/h4-8H,3H2,1-2H3,(H,17,19)
Lösningstyp Neat
LEDER CCN1c2nccc(C)c2NC(=O)c3cccnc13
IUPAC-namn 11-ethyl-4-methyl-5H-dipyrido[2,3-b:3',2'-e][1,4]diazepin-6-one
Molekylvikt (g/mol) 254.29
Kvantitet 4 x 25 mg
Formel vikt 254.1168 g/mol