Läs mer
Nexturastat A, MedChemExpress
Produktkod. 16487138
Encompass_Preferred
Change view
Kvantitet:
1 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
1mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16487138 10 mg -
16481629 1 mg 1mg
16463167 50 mg -
16437146 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16487138 Leverantör Medchem Express Leverantörsnummer HY16699
MedChemExpress Nexturastat A is a potent HDAC6 inhibitor with an IC50 of 5 nM. Nexturastat A also inhibits HDAC10 and metallo-β-lactamase domain-containing protein 2 (MBLAC2).
Nexturastat A is a potent HDAC6 inhibitor with an IC50 of 5 nM. Nexturastat A also inhibits HDAC10 and metallo-β-lactamase domain-containing protein 2 (MBLAC2).
Specifikationer
Kemiskt namn eller material Nexturastat A
Fysisk form Solid
CAS 1403783-31-2
Färg White
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C19H23N3O3
För användning med (applikation) Cancer-programmed cell death
Löslighetsinformation DMSO : ≥ 56 mg/mL (164.03 mM)
LEDER O=C(NO)C1=CC=C(CN(CCCC)C(NC2=CC=CC=C2)=O)C=C1
Molekylvikt (g/mol) 341.4
Formel vikt 341.4
Procent renhet 99.03%