missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ NFκB/p65 (Rel A) Ab-1 Rabbit Polyclonal Antibody with BSA and azide

Epredia™ NFκB/p65 (Rel A) Ab-1, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Brand:  Epredia™ RB-1638-P0

Additional Details : Weight : 0.00170kg

 View more versions of this product

Product Code. 12643097

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

NFκ B is a heterodimer consisting of a 50kDa DNA binding subunit and a 65kDa transactivating subunit (RelA). The p50/p65 heterodimer is present in the cytoplasm in inactive form bound to its inhibitor, IdeltaB. Upon stimulation of cells with stimuli like LPS, Interleukin-1, IkB gets phosphorylated and degraded by proteosomes releasing the activated NFκB which goes to the nucleus inducing gene expression. NFκB was originally identified as transcriptional activator for kappa light chain gene in B cells.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat, Hamster, Rabbit, Cow, Pig, Dog, Drosophila, Xenopus. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminus of human NFκB protein

Molecular Weight: 65kDa

Positive Control: Jurkat cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

NFkappaB/p65 (Rel A) Ab-1
Polyclonal
Unconjugated
A synthetic peptide from the C-terminus of human NFkappaB protein
RUO
Primary
Concentrated
Western Blot
1mg/mL
Rabbit
100 μL
Transcription Regulation
Bovine, Canine, Drosophila, Hamster, Human, Mouse, Porcine, Rabbit, Rat, Xenopus species
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers