missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF2/Merlin Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99421R
3212.00 SEK valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
NF2/Merlin Polyclonal antibody specifically detects NF2/Merlin in Mouse samples. It is validated for Western Blot
Specifications
NF2/Merlin | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot | |
DyLight 550 | |
ACN, BANF, merlin, moesin-ezrin-radixin like, Moesin-ezrin-radixin-like protein, moesin-ezrin-radizin-like protein, neurofibromin 2 (bilateral acoustic neuroma), neurofibromin 2 (merlin), neurofibromin-2, SCH, Schwannomerlin, Schwannomin | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human NF2/Merlin | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Tumor Suppressors | |
4771 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction