Läs mer
Tocris Bioscience™ NFAT inhibitor, Cell Permeable
Cell permeable NFAT inhibitor
Brand: Tocris Bioscience™ 5710/1
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
NFAT inhibitor, Cell Permeable is a NFAT inhibitor. Inhibits LPS or LPS plus IFN-γ-induced IL-12 p40, IL-12 p70, IL-23 and TNF secretion from bone marrow-derived macrophages (BMDMs). Also attenuates NO production and Nos2 mRNA expression in LPS-stimulated BMDMs. Improves symptoms in a mouse model of colitis. Exhibits immunosuppressive effects; enhances graft survival in mice. Cell permeable.Specifikationer
| 592517-80-1 | |
| Protein Ser/Thr Phosphatase Inhibitors | |
| 1 mg |
| Store at -20°C | |
| Observed M.W.: 3573.15 | |
| C147H259N67O36S |