missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFE2L1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | NFE2L1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16876884
|
Proteintech
17062-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16866884
|
Proteintech
17062-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human ESRRG.
This gene encodes a protein that is involved in globin gene expression in erythrocytes. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene, NFE2L1, and for ′nuclear respiratory factor 1′ which has an official symbol of NRF1.Specifications
NFE2L1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q14494, Q61985 | |
18023, 4779 | |
NFE2L1 Fusion Protein Ag10832 | |
Primary | |
-20°C | |
NFE2L1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HBZ17, LCR F1, Locus control region factor 1, NF E2 related factor 1, NFE2 related factor 1, NFE2L1, NRF1, TCF 11, TCF11, Transcription factor 11, Transcription factor HBZ17, Transcription factor LCR F1 | |
Nfe2l1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |