missing translation for 'onlineSavingsMsg'
Läs mer

NFkB p105/p50 Antibody (2J10D7), DyLight 680, Novus Biologicals™

Produktkod. 18075084 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18075084 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18075084 Leverantör Novus Biologicals Leverantörsnummer NB10056583FR

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

NFkB p105/p50 Monoclonal specifically detects NFkB p105/p50 in Human, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen NFkB p105/p50
Användningsområden Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
Klassificering Monoclonal
Klona 2J10D7
Konjugera DyLight 680
Utspädning Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated
Genaccessionsnr. P25942
Gene Alias DKFZp686C01211, DNA binding factor KBF1, DNA-binding factor KBF1, EBP-1, KBF1, NF-kappaB, NF-kappa-B, NF-kappabeta, NFKB-p105, NFKB-p50, nuclear factor kappa-B DNA binding subunit, nuclear factor NF-kappa-B p105 subunit, nuclear factor NF-kappa-B p50 subunit, nuclear factor of kappa light polypeptide gene enhancer in B-cells 1MGC54151, p105, p50
Gensymboler NFKB1
Värd art Mouse
Immunogen A portion of amino acids 150-200 of human NF-kB (p50) was used as the immunogen.
Reningsmetod Protein G purified
Kvantitet 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Apoptosis, Cancer, Cell Biology, Diabetes Research, DNA Repair, Neurodegeneration, Neuroscience, Nucleotide Excision Repair, Signal Transduction, Transcription Factors and Regulators
Primär eller sekundär Primary
Gen-ID (Entrez) 4790
Testspecificitet Cross reacts with Human. Predicted to react with Mouse and Rat which are 86% homologous with the immunogen.
Målarter Human, Rat, Rabbit
Innehåll och lagring Store at 4C in the dark.
Form Purified
Isotyp IgG1 κ
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.