missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFKB1 p105/p50 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15506-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody can bind both p105 and p50 isoforms of NFKB1.
NF-kB is a nuclear transcription factor activated by various extra and intracellular stimuli such as cytokines, UV radiation, stress, in injury, and by bacterial and viral products. It is involved in regulation of various cellular events including cell growth, differentiation, proliferation, apoptosis and inflammation. NFKB1 (p50), a 50 kDa functional sub-unit of NF-kB, is a member of Rel protein family. It is synthesized as a p105 precursor protein and consists of an N-terminal conserved RHD-region containing nuclear localization signal, DNA-binding and dimerization domains. NFKB1 (p50) forms homodimers or heterodimerizes with p65, forming the functional NF-kB factor. NFKB1 (p50) directs the nuclear translocation of NF-kB and is instrumental in its DNA-binding. Pathological role of NF-kB has been suggested in AIDS, hematogenic cancer cell metastasis and rheumatoid arthritis.Specifications
NFKB1 p105/p50 | |
Polyclonal | |
Unconjugated | |
NFKB1 | |
DNA binding factor KBF1, EBP 1, KBF1, NF kappa B, NFKB p105, NFKB p50, NFKB1, NFKB1,p105, NFKB1,p105,p50, p105, p50 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4790 | |
-20°C | |
Liquid |
Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P19838 | |
NFKB1 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |