missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NFKBIA (Human) Recombinant Protein
Human NFKBIA full-length ORF ( AAH04983, 1 a.a. - 317 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004792-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MFQAAERPQEWAMEGPRDGLKKERLLDDRHDSGLDSMKDEEYEQMVKELQEIRLEPQEVPRGSEPWKQQLTEDGDSFLHLAIIHEEKALTMEVIRQVKGDLAFLNFQNNLQQTPLHLAVITNQPEIAEALLGAGCDPELRDFRGNTPLHLACEQGCLASVGVLTQSCTTPHLHSILKATNYNGHTCLHLASIHGYLGIVELLVSLGADVNAQEPCNGRTALHLAVDLQNPDLVSLLLKCGADVNRVTYQGYSPYQLTWGRPSTRIQQQLGQLTLENLQMLPESEDEESYDTESEFTEFTEDELPYDDCVFGGQRLTL
Specifications
AAH04983 | |
nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NFKBIA | |
GST |
4792 | |
Wheat germ expression system | |
10 μg | |
IKBA, MAD-3, NFKBI | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |