Läs mer
Tocris Bioscience™ NFPS
GREENER_CHOICE
Produktkod. 18025662
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18025662 50 mg
18004172 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18025662 Leverantör Tocris Bioscience™ Leverantörsnummer 2789/50
Selective non-transportable GlyT1 inhibitor
Selective, non-transportable inhibitor of GlyT1 which displays no activity at GABA or glutamate receptors (IC50 values are 2.8, 9.8 and 56000 nM for hGlyT1, rGlyT1 and rGlyT2 respectively). Enhances the amplitude of the NMDA component of glutamatergic EPSCs.
R-enantiomer also available.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 405225-21-0
Molekylformel C24H24FNO3
Specifikationer
Kemiskt namn eller material NFPS
Kvantitet 50 mg
Formel vikt 393.45
Renhet >98%
Molekylvikt (g/mol) 393.458