missing translation for 'onlineSavingsMsg'
Learn More

NFU1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25598-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824165

  • 1236.20 SEK / 20µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein that is localized to mitochondria and plays a critical role in iron-sulfur cluster biogenesis. The encoded protein assembles and transfers 4Fe-4S clusters to target apoproteins including succinate dehydrogenase and lipoic acid synthase. Mutations in this gene are a cause of multiple mitochondrial dysfunctions syndrome-1, and pseudogenes of this gene are located on the short arms of chromosomes 1 and 3. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications

Specifications

NFU1
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9QZ23, Q9UMS0
CGI 33, HIRA interacting protein 5, HIRIP5, Nfu, NFU1, NifU, NIFUC
Rabbit
IgG
20 μL
-20°C
Primary
27247, 297416, 56748
Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry
Unconjugated
NFU1
Liquid
Nfu1
NFU1 Fusion Protein Ag22343
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers