missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NFYB (Human) Recombinant Protein
Human NFYB full-length ORF ( AAH05317, 1 a.a. - 207 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004801-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MTMDGDSSTTDASQLGISADYIGGSHYVIQPHDDTEDSMNDHEDTNGSKESFREQDIYLPIANVARIMKNAIPQTGKIAKDAKECVQECVSEFISFITSEASERCHQEKRKTINGEDILFAMSTLGFDSYVEPLKLYLQKFREAMKGEKGIGGAVTATDGLSEELTEEAFTNQLPAGLITTDGQQQNVMVYTTSYQQISGVQQIQFS
Specifications
AAH05317 | |
nuclear transcription factor Y, beta | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NFYB | |
GST |
4801 | |
Wheat germ expression system | |
25 μg | |
CBF-A, CBF-B, HAP3, NF-YB | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |