Läs mer
Tocris Bioscience™ NG 25
TGF-β-activated kinase (TAK1) inhibitor
Brand: Tocris Bioscience™ 5429/5
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
NG 25 is a TGF-β-activated kinase (TAK1) inhibitor (IC50 = 149 nM). Also inhibits LYN, MAP4K2 and Abl kinase activity (IC50 values are 12.9, 21.7 and 75.2 nM, respectively). Suppresses CpG A and CpG B-stimulated secretion of IFN-α and TLR7/8 agonist CL097-stimulated secretion of IFN-β in Gen 2.2 leukemia cells in vitro.Specifikationer
| N-[4-[(4-Ethyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]-4-methyl-3-(1H-pyrrolo[2,3-b]pyridin-4-yloxy)benzamide | |
| 1315355-93-1 | |
| 5 mg |