Läs mer
NG,NG-Dimethylarginine Dihydrochloride, TRC
Produktkod. 17991835
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17991835 5 mg
17964775 10 mg
17931405 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17991835 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4635755MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 220805-22-1
Molekylformel C8 H18 N4 O2 . 2 Cl H
Molekylvikt (g/mol) 275.18
Synonym L-Ornithine, N5-[(dimethylamino)iminomethyl]-, hydrochloride (1:2), L-Ornithine, N5-[(dimethylamino)iminomethyl]-, dihydrochloride (9CI), N5-[(Dimethylamino)iminomethyl]-L-ornithine dihydrochloride, (2S)-2-Amino-5-[[amino(dimethylamino)methylidene]amino]pentanoic acid dihydrochloride, (S)-2-Amino-5-(3,3-dimethylguanidino)pentanoic acid dihydrochloride, ADMA 2HCl, Asymmetric dimethylarginine dihydrochloride, Dimethyl-L-arginine dihydrochloride, L-NG,NG-Dimethylarginine dihydrochloride, NG,NG-Dimethylarginine dihydrochloride, NG,NG-Dimethyl-L-arginine dihydrochloride
IUPAC-namn (2S)-2-amino-5-[(N,N-dimethylcarbamimidoyl)amino]pentanoic acid;dihydrochloride
LEDER Cl.Cl.CN(C)C(=N)NCCC[C@H](N)C(=O)O
Specifikationer
Kemiskt namn eller material NG,NG-Dimethylarginine Dihydrochloride
InChI formel InChI=1S/C8H18N4O2.2ClH/c1-12(2)8(10)11-5-3-4-6(9)7(13)14;;/h6H,3-5,9H2,1-2H3,(H2,10,11)(H,13,14);2*1H/t6-;;/m0../s1
Kvantitet 5 mg
Formel vikt 274.0963