Läs mer
Invitrogen™ NG2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582046
Beskrivning
Immunogen sequence: ILYEHEMPPE PFWEAHDTLE LQLSSPPARD VAATLAVAVS FEAACPQRPS HLWKNKGLWV PEGQRARITV AALDASNLLA SVPSPQRSEH DVLFQVTQFP SRGQLLVSEE PLHAGQPHFL QSQLAAGQLV YAHGGGGTQQ DGFHFRAHL Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 83%.
A human melanoma-associated chondroitin sulfate proteoglycan plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes. CSPG4 represents an integral membrane chondroitin sulfate proteoglycan expressed by human malignant melanoma cells.
Specifikationer
| NG2 | |
| Polyclonal | |
| Unconjugated | |
| Cspg4 | |
| 4732461B14Rik; AN2; AN2 proteoglycan; AN2/NG2 proteoglycan; chondroitin sulfate proteoglycan 4; chondroitin sulfate proteoglycan 4 (melanoma-associated); chondroitin sulfate proteoglycan NG2; CSPG4; HMW MAA; HMW-MAA; HSN tumor-specific antigen; Kiaa4232; MCSP; MCSPG; MEL CSPG; melanoma chondroitin sulfate proteoglycan; Melanoma-associated chondroitin sulfate proteoglycan; MEL-CSPG; membrane-spanning proteoglycan NG2; MSK16; nerve/glial antigen 2; Ng2; Proteoglycan AN2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1464 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UVK1 | |
| Cspg4 | |
| Recombinant protein corresponding to Human CSPG4. Recombinant protein control fragment (Product #RP-102032). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.