Learn More
Invitrogen™ NGFR Human ELISA Kit
Sandwich ELISA Kit
5870.00 SEK - 27960.00 SEK
Specifications
Accession Number | P08138 |
---|---|
Assay Sensitivity | 80 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15435083
|
Invitrogen™
EHNGFR |
96 Tests |
5870.00 SEK
1 set |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17155519
|
Invitrogen™
EHNGFRX5 |
5 x 96 Tests |
27960.00 SEK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
The Human Nerve Growth Factor Receptor (Hu NGFR) ELISA quantitates Hu NGFR in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu NGFR. Principle of the method The Human NGFR solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
NGFR (Nerve Growth Factor Receptor, TNFR super family, member 16, LNGFR, p75 neurotrophin receptor) is important for the development, differentiation, and survival of variety of neuronal and non-neuronal cells. NGF action is mediated by binding two distinct receptors, the high affinity p140 and low affinity Nerve Growth Factor (NGF) Receptor (p75NGFR) or Neurotrophin Receptor (p75NTR). NGFR contains an extracellular domain containing four 40-amino acid repeats with 6 cysteine residues at conserved positions followed by a serine/threonine-rich region, a single transmembrane domain, and a 155-amino acid cytoplasmic domain. The cysteine-rich region contains the nerve growth factor binding domain. Neurotrophic factors control the survival, differentiation and maintenance of neurons in the peripheral and central nervous systems, and of other neural crest-derived cell types. Developing sympathetic neurons are absolutely dependent upon nerve growth factor during the period of target competition in vivo. Antibodies reacting specifically with NGFR p75 are useful tools in the detection and characterization of NGFR p75, to enhance our understanding of a wide range of phenomena in the development, plasticity and repair of the nervous system. NGFR can also associate with other NGF receptors such as Trk through the cytosolic and transmembrane domains and thus can function as a co-receptor that refines Trk affinity and specificity for neurotrophins. Finally, upon binding of various neurotrophins, NGFR associates with tumor necrosis factor receptor-6 (TRAF6), suggesting that it can potentially function as a signal transducer for NGF signals through NGFR. NGFR may play a role in the development of cancer stem cells in melanoma and other cancer types.Specifications
P08138 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Human | |
Colorimetric Microplate Reader | |
CD271,Gp80-LNGFR,TNFRSF16,p75(NTR),p75NTR | |
<10% | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
80 pg/mL | |
Biotin | |
Growth Factors & Receptors | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF- 4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
4804 | |
<12% | |
HRP | |
Apoptosis | |
CD271, Gp80-LNGFR, TNFRSF16, p75(NTR), p75NTR | |
1 hr. 20 min. |