missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
NHERF-1 Antibody (EBP-10), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-488V
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
NHERF-1 Monoclonal antibody specifically detects NHERF-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifikationer
| NHERF-1 | |
| Monoclonal | |
| DyLight 405 | |
| EBP50NHERF-1, ezrin-radixin-moesin binding phosphoprotein-50, Ezrin-radixin-moesin-binding phosphoprotein 50, Na(+)/H(+) exchange regulatory cofactor NHE-RF1, Na+/H+ exchange regulatory co-factor, NHERF1solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 1, NHERFNPHLOP2, Regulatory cofactor of Na(+)/H(+) exchanger, sodium/hydrogen exchanger regulatory factor 1, Sodium-hydrogen exchanger regulatory factor 1, solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatoryfactor 1, solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 1, Solute carrier family 9 isoform A3 regulatory factor 1 | |
| Bacterially produced recombinant full-length human NHERF1. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Signal Transduction | |
| 9368 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| EBP-10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Spot an opportunity for improvement?Share a Content Correction