Läs mer
Nicergoline EP Impurity C, Mikromol™
Produktkod. 17923416
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17923416 25 mg
17943906 100 mg
30463462 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17923416 Leverantör Mikromol Leverantörsnummer MM0115.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Nicergoline EP Impurity C
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methylmethoxylumilysergol
CAS 35155-28-3
Analyt- eller komponentnamn Methylmethoxylumilysergol
Frakt skick Room Temperature
Molekylformel C18 H24 N2 O2
Certifikat och efterlevnad ISO 9001
Synonym [(6aR,9R,10aS)-10a-Methoxy-4,7-dimethyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinolin-9-yl]methanol (Methylmethoxylumilysergol), [(6aR,9R,10aS)-10a-Methoxy-4,7-dimethyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinolin-9-yl]methanol, Nicergoline Imp. C (EP), Methylmethoxylumilysergol, Ergoline-8-methanol, 10-methoxy-1,6-dimethyl-, (8beta)-, Lumilysergol, 10O,1-dimethyl- (7CI), (8beta)-10-Methoxy-1,6-dimethylergoline-8-methanol, Indolo[4,3-fg]quinoline, ergoline-8-methanol deriv., (+)-(5R,10S)-Meluol, 1,6-Dimethyl-8beta-(hydroxymethyl)-10alpha-methoxyergoline, 1-Methyl-10alpha-methoxydihydrolysergol, 10-Methoxy-1,6-dimethylergoline-8beta-methanol, 10alpha-Methoxy-1-methyl-9,10-dihydrolysergol, 10alpha-Methoxy-1-methyldihydrolysergol, O10,1-Dimethyllumilysergol, Nicergoline Impurity C
InChI formel InChI=1S/C18H24N2O2/c1-19-10-13-7-16-18(22-3,8-12(11-21)9-20(16)2)14-5-4-6-15(19)17(13)14/h4-6,10,12,16,21H,7-9,11H2,1-3H3/t12-,16-,18+/m1/s1
LEDER CO[C@]12C[C@@H](CO)CN(C)[C@@H]1Cc3cn(C)c4cccc2c34
IUPAC-namn [(6aR,9R,10aS)-10a-methoxy-4,7-dimethyl-6a,8,9,10-tetrahydro-6H-indolo[4,3-fg]quinoline-9-yl]methanol
Molekylvikt (g/mol) 300.4
Kvantitet 25 mg
Formel vikt 300.1838 g/mol