Learn More
Nickelous Sulfate, 6-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Catalog Number p-5390799
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
10101-97-0
H12NiO10S
262.839
RRIWRJBSCGCBID-UHFFFAOYSA-L
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
5284429
CHEBI:53437
nickel(2+);sulfate;hexahydrate
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
10101-97-0
H12NiO10S
262.839
RRIWRJBSCGCBID-UHFFFAOYSA-L
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
5284429
CHEBI:53437
nickel(2+);sulfate;hexahydrate
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
High quality chemicals for laboratory and specialized industrial use
Physical Form Crystals
Quantity 500 g
Formula Weight 262.85
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.03kg
Chemical Name or Material Nickelous Sulfate, 6-Hydrate
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H351-Suspected of causing cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.