missing translation for 'onlineSavingsMsg'
Läs mer

Nicosulfuron-d6, TRC

Product Code. 30473880
Change view
Click to view available options
Kvantitet:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30473880 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30473880 Supplier Toronto Research Chemicals Supplier No. TRCN3995021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Nicosulfuron-d6
CAS 1189419-41-7
Anteckningar om renhetsgrad HPLC
Molekylformel C15 2H6 H12 N6 O6 S
Kvantitet 1 mg
Synonym Nicosulfuron D6 (Dimethoxy D6), 2-[[4,6-Bis(trideuteriomethoxy)pyrimidin-2-yl]carbamoylsulfamoyl]-N,N-dimethylpyridine-3-carboxamide, Nicosulfuron D6 (Dimethoxy D6), Nicosulfuron D6 (dimethoxy D6), 2-[[4,6-Bis(trideuteriomethoxy)pyrimidin-2-yl]carbamoylsulfamoyl]-N,N-dimethylpyridine-3-carboxamide
InChI formel InChI=1S/C15H18N6O6S/c1-21(2)13(22)9-6-5-7-16-12(9)28(24,25)20-15(23)19-14-17-10(26-3)8-11(18-14)27-4/h5-8H,1-4H3,(H2,17,18,19,20,23)/i3D3,4D3
LEDER [2H]C([2H])([2H])Oc1cc(OC([2H])([2H])[2H])nc(NC(=O)NS(=O)(=O)c2ncccc2C(=O)N(C)C)n1
IUPAC-namn 2-[[4,6-bis(trideuteriomethoxy)pyrimidin-2-yl]carbamoylsulfamoyl]-N,N-dimethylpyridine-3-carboxamide
Molekylvikt (g/mol) 416.44
Formel vikt 416.1385
Procent renhet >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.