Läs mer
Nicotianamine, TRC
Produktkod. 30012914
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012914 10 mg
30009817 5 mg
30008504 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012914 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN40850010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34441-14-0
Molekylformel C11 H19 N3 O6
Molekylvikt (g/mol) 289.29
IUPAC-namn (2S)-1-[(3S)-3-[[(3S)-3-amino-3-carboxypropyl]amino]-3-carboxypropyl]aziridine-2-carboxylic acid
LEDER N[C@@H](CCN[C@@H](CCN1C[C@H]1C(=O)O)C(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material Nicotianamine
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C11H19N3O6/c12-6(9(15)16)1-3-13-7(10(17)18)2-4-14-5-8(14)11(19)20/h6-8,13H,1-5,12H2,(H,15,16)(H,17,18)(H,19,20)/t6-,7-,8-,14?/m0/s1
Formel vikt 289.1274
Procent renhet >95%