Läs mer
Nicotinamide 1,N6-Ethenoadenine Dinucleotide, TRC
Produktkod. 30011830
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011830 5 mg
30008579 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30011830 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN4077355MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 38806-38-1
Molekylformel C23 H27 N7 O14 P2
Molekylvikt (g/mol) 687.4465
Synonym Pyridinium, 3-(aminocarbonyl)-1-[5-O-[hydroxy(phosphonooxy)phosphinyl]-β-D-ribofuranosyl]-, inner salt, P'→5'-ester with 3-β-D-ribofuranosyl-3H-imidazo[2,1-i]purine, 3H-Imidazo[2,1-i]purine, pyridinium deriv., 1,N6-Ethenonicotinamide adenine dinucleotide, ε-NAD, 3-(Aminocarbonyl)-1-[5-O-[hydroxy(phosphonooxy)phosphinyl]-β-D-ribofuranosyl]pyridinium Inner Salt P'5'-Ester with 3-β-D-Ribofuranosyl-3H-imidazo[2,1-i]purine, 1,N6-Ethenonicotinamide Adenine Dinucleotide
IUPAC-namn [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl [[(2R,3S,4R,5R)-3,4-dihydroxy-5-imidazo[2,1-f]purin-3-yloxolan-2-yl]methoxy-hydroxyphosphoryl] phosphate
LEDER NC(=O)c1ccc[n+](c1)[C@@H]2O[C@H](COP(=O)([O-])OP(=O)(O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)n4cnc5c4ncn6ccnc56)[C@@H](O)[C@H]2O
Specifikationer
Kemiskt namn eller material Nicotinamide 1,N6-Ethenoadenine Dinucleotide
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C23H27N7O14P2/c24-19(35)11-2-1-4-28(6-11)22-17(33)15(31)12(42-22)7-40-45(36,37)44-46(38,39)41-8-13-16(32)18(34)23(43-13)30-10-26-14-20-25-3-5-29(20)9-27-21(14)30/h1-6,9-10,12-13,15-18,22-23,31-34H,7-8H2,(H3-,24,35,36,37,38,39)/t12-,13-,15-,16-,17-,18-,22-,23-/m1/s1
Formel vikt 687.1091
Procent renhet >95%