missing translation for 'onlineSavingsMsg'
Learn More

β-Nicotinamide Adenine Dinucleotide Phosphate, Monosodium Salt, Tetrahydrate, Thermo Scientific Chemicals

Catalog Number p-7082800
Click to view available options
Quantity:
250 mg
500 mg
1 g
5 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
1184-16-3
C21H27N7NaO17P3
765.39
MFCD00036973
JNUMDLCHLVUHFS-QYZPTAICSA-M
nadp sodium salt, beta-nicotinamide adenine dinucleotide phosphate sodium
131845262
[[(2 R,3 R,4 R,5 R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2 R,3 S,4 R,5 R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodiμm
[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
This item is not returnable. View return policy
1184-16-3
C21H27N7NaO17P3
765.39
MFCD00036973
JNUMDLCHLVUHFS-QYZPTAICSA-M
nadp sodium salt, beta-nicotinamide adenine dinucleotide phosphate sodium
131845262
[[(2 R,3 R,4 R,5 R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2 R,3 S,4 R,5 R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodiμm
[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
Physical Form Powder
Quantity 250 mg
Beilstein 4779954
Formula Weight 837.45
Packaging Plastic bottle
Chemical Name or Material beta-Nicotinamide adenine dinucleotide phosphate monosodium salt tetrahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'